accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CCH3NH2HCl

CAS RN 23358-42-1

IUPAC Name: ethyl 2-amino-3-methylbutanoate hydrochloride
Synonyms: ethyl 2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: PQGVTLQEKCJXKF-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)C(N)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)