accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeOHOOOOOO

CAS RN 2338-05-8

IUPAC Name: iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: ferric citrate
Molecular Weight (g/mol): 244.94
Molecular Formula: C6H5FeO7
InChi Key: NPFOYSMITVOQOS-UHFFFAOYSA-K
SMILES: [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Grade 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)