accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOOHN

CAS RN 2338-19-4

IUPAC Name: potassium 2-(1H-indol-3-yl)acetate
Synonyms: potassium β-indoleacetate
Molecular Weight (g/mol): 213.28
Molecular Formula: C10H8KNO2
InChi Key: MLWMEUAQPRACHM-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results