accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2338-71-8

FHNO

CAS RN 2338-71-8

IUPAC Name: 5-fluoro-1H-indole-3-carbaldehyde
Synonyms: 5-fluoro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: YUAJKGBLPVLADK-UHFFFAOYSA-N
SMILES: FC1=CC=C2NC=C(C=O)C2=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results