accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNSO

CAS RN 23417-29-0

IUPAC Name: 6-methyl-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 6-methyl-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: NUMDWMDMUXZMHA-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=S)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results