accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3HNOONNCH3H3C(E/Z)

CAS RN 23422-53-9

IUPAC Name: hydrogen 3-{[(dimethylamino)methylidene]amino}phenyl N-methylcarbamate chloride
Synonyms: formetanate
Molecular Weight (g/mol): 257.72
Molecular Formula: C11H16ClN3O2
InChi Key: MYPKGPZHHQEODQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNC(=O)OC1=CC=CC(=C1)N=CN(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results