CAS RN 23431-48-3
CAS RN 23431-48-3
IUPAC Name:
1-methyl-4-(prop-2-en-1-yloxy)benzene
Synonyms:
1-methyl-4-(prop-2-en-1-yloxy)benzene
Molecular Weight (g/mol):
148.21
Molecular Formula:
C10H12O
InChi Key:
AROCNZZBLCAOPH-UHFFFAOYSA-N
SMILES:
CC1=CC=C(OCC=C)C=C1
1
–
1
of
1
results
1
Allyl p-Tolyl Ether 95.0+%, TCI America™
CAS: 23431-48-3 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.205 MDL Number: MFCD00026098 InChI Key: AROCNZZBLCAOPH-UHFFFAOYSA-N Synonym: 4-Allyloxytoluene PubChem CID: 262070 IUPAC Name: 1-methyl-4-prop-2-enoxybenzene SMILES: CC1=CC=C(C=C1)OCC=C