accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNO

CAS RN 23432-43-1

IUPAC Name: 6-chloro-1,4-dihydroquinolin-4-one
Synonyms: 6-chloro-1H-quinolin-4-one
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: XXGUQCVVGPZTPF-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC=CC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results