accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2345-27-9

H3CCH3O

CAS RN 2345-27-9

IUPAC Name: tetradecan-2-one
Synonyms: dodecyl methyl ketone
Molecular Weight (g/mol): 212.38
Molecular Formula: C14H28O
InChi Key: POQLVOYRGNFGRM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)