accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrBrCH3BrBr

CAS RN 23488-38-2

IUPAC Name: 1,2,4,5-tetrabromo-3,6-dimethylbenzene
Synonyms: 1,2,4,5-tetrabromo-3,6-dimethylbenzene
Molecular Weight (g/mol): 421.75
Molecular Formula: C8H6Br4
InChi Key: RXKOKVQKECXYOT-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(Br)=C(C)C(Br)=C1Br
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)