accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH(S)NHOOH3CH3CCH3OOH

CAS RN 23500-04-1

IUPAC Name: 6-{[(tert-butoxy)carbonyl]amino}-2-acetamidohexanoic acid
Synonyms: 6-[(tert-butoxycarbonyl)amino]-2-acetamidohexanoic acid
Molecular Weight (g/mol): 288.34
Molecular Formula: C13H24N2O5
InChi Key: NZAMQYCOJSWUAO-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)