accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OCH3NNNH3CH3C

CAS RN 23505-41-1

IUPAC Name: O-2-(diethylamino)-6-methylpyrimidin-4-yl O,O-diethyl phosphorothioate
Synonyms: pirimiphos ethyl
Molecular Weight (g/mol): 333.39
Molecular Formula: C13H24N3O3PS
InChi Key: TZBPRYIIJAJUOY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CC(C)=NC(=N1)N(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results