CAS RN 235088-69-4
CAS RN 235088-69-4
IUPAC Name:
1-(3,4,5-trifluorophenyl)methanamine
Synonyms:
1-(3,4,5-trifluorophenyl)methanamine
Molecular Weight (g/mol):
161.13
Molecular Formula:
C7H6F3N
InChi Key:
AJGBQAAXUVSBCH-UHFFFAOYSA-N
SMILES:
NCC1=CC(F)=C(F)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 3,4,5-Trifluorobenzylamine
Sigma Aldrich 3,4,5-Trifluorobenzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.