accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 235099-48-6

IUPAC Name: 9,10-bis(6-methoxynaphthalen-2-yl)anthracene
Synonyms: 9,10-bis(6-methoxynaphthalen-2-yl)anthracene
Molecular Weight (g/mol): 490.6
Molecular Formula: C36H26O2
InChi Key: VEVBQRGODPIBNJ-UHFFFAOYSA-N
SMILES: COC1=CC2=CC=C(C=C2C=C1)C1=C2C=CC=CC2=C(C2=CC3=CC=C(OC)C=C3C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)