accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23528-44-1

IUPAC Name: {[2-(phenylsulfanyl)ethenyl]sulfanyl}benzene
Synonyms: {[2-(phenylsulfanyl)ethenyl]sulfanyl}benzene
Molecular Weight (g/mol): 244.37
Molecular Formula: C14H12S2
InChi Key: UHGSEDVQATYWGI-UHFFFAOYSA-N
SMILES: S(C=CSC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results