accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOCH2(E)

CAS RN 23578-51-0

IUPAC Name: (2E)-octa-2,7-dien-1-ol
Synonyms: 2,7-octadien-1-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: YHYGSIBXYYKYFB-VOTSOKGWSA-N
SMILES: OC\C=C\CCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results