accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23586-53-0

IUPAC Name: thallium(3+) tritrifluoroacetate
Synonyms: thallium(3+) tritrifluoroacetate
Molecular Weight (g/mol): 543.43
Molecular Formula: C6F9O6Tl
InChi Key: PSHNNUKOUQCMSG-UHFFFAOYSA-K
SMILES: [Tl+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)