accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHOOHO

CAS RN 2359-09-3

IUPAC Name: 5-tert-butylbenzene-1,3-dicarboxylic acid
Synonyms: 5-tert-butylbenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: BJLUCDZIWWSFIB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1)C(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results