accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23598-72-3

IUPAC Name: 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 237.64
Molecular Formula: C11H8ClNO3
InChi Key: UVEPOHNXGXVOJE-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)