accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2360-97-6

OOOHNH2NN(S)

CAS RN 2360-97-6

IUPAC Name: (2S)-2-amino-N-(4-nitrophenyl)-3-phenylpropanamide
Synonyms: (2S)-2-amino-N-(4-nitrophenyl)-3-phenylpropanamide
Molecular Weight (g/mol): 285.3
Molecular Formula: C15H15N3O3
InChi Key: GJHIOWXZFDVUKQ-AWEZNQCLSA-N
SMILES: N[C@@H](CC1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)