accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)(S)

CAS RN 2362-61-0

IUPAC Name: (1R,2S)-2-phenylcyclohexan-1-ol
Synonyms: (1R,2S)-2-phenylcyclohexan-1-ol
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: AAIBYZBZXNWTPP-NWDGAFQWSA-N
SMILES: O[C@@H]1CCCC[C@H]1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results