accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 2363-71-5

IUPAC Name: henicosanoic acid
Synonyms: heneicosanoic acid
Molecular Weight (g/mol): 326.57
Molecular Formula: C21H42O2
InChi Key: CKDDRHZIAZRDBW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCC(O)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)