accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2(S)OO

CAS RN 23635-14-5

IUPAC Name: (4S)-4-(prop-1-en-2-yl)cyclohex-1-ene-1-carboxylate
Synonyms: perillic acid
Molecular Weight (g/mol): 165.21
Molecular Formula: C10H13O2
InChi Key: CDSMSBUVCWHORP-MRVPVSSYSA-M
SMILES: CC(=C)[C@H]1CCC(=CC1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results