accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHOHHOHOO

CAS RN 23651-95-8

IUPAC Name: 2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid
Synonyms: droxidopa
Molecular Weight (g/mol): 213.19
Molecular Formula: C9H11NO5
InChi Key: QXWYKJLNLSIPIN-UHFFFAOYNA-N
SMILES: NC(C(O)C1=CC=C(O)C(O)=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)