accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHFF

CAS RN 2366-62-3

IUPAC Name: 2,2-difluorobutanoic acid
Synonyms: 2,2-difluorobutanoic acid
Molecular Weight (g/mol): 124.09
Molecular Formula: C4H6F2O2
InChi Key: SBSKRVZFWQPJID-UHFFFAOYSA-N
SMILES: CCC(F)(F)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)