accessibility menu, dialog, popup

UserName

Viewing profile as user:
CZnI

CAS RN 23665-09-0

IUPAC Name: benzenide; iodozincylium
Synonyms: benzenide; iodozincylium
Molecular Weight (g/mol): 269.39
Molecular Formula: C6H5IZn
InChi Key: QXXJGZQLPDQOPF-UHFFFAOYSA-M
SMILES: [Zn+]I.C1=CC=[C-]C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results