accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23676-09-7

IUPAC Name: ethyl 4-ethoxybenzoate
Synonyms: ethyl 4-ethoxybenzoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: HRAQMGWTPNOILP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(OCC)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)