accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23680-31-1

IUPAC Name: 3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: DMBKPDOAQVGTST-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)