accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2369-37-1

IUPAC Name: 1,2-dibromo-4-fluorobenzene
Synonyms: 1,2-dibromo-4-fluorobenzene
Molecular Weight (g/mol): 253.9
Molecular Formula: C6H3Br2F
InChi Key: RNTGKISRXVFIIP-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)