accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiOSiCH3OSiCH3OSiH3CO

CAS RN 2370-88-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 236.48
Molecular Formula: C4H12O4Si4
InChi Key: WZJUBBHODHNQPW-UHFFFAOYSA-N
SMILES: C[Si]1O[Si](C)O[Si](C)O[Si](C)O1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)