accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23702-54-7

IUPAC Name: 4-{[(3,4-dimethoxyphenyl)methoxy]methyl}-1,2-dimethoxybenzene
Synonyms: 4-{[(3,4-dimethoxyphenyl)methoxy]methyl}-1,2-dimethoxybenzene
Molecular Weight (g/mol): 318.37
Molecular Formula: C18H22O5
InChi Key: KTFBMMKWTQVUIV-UHFFFAOYSA-N
SMILES: COC1=CC=C(COCC2=CC=C(OC)C(OC)=C2)C=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results