accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2373-80-0

IUPAC Name: (2Z)-3-(2H-1,3-benzodioxol-5-yl)prop-2-enoic acid
Synonyms: (2Z)-3-(2H-1,3-benzodioxol-5-yl)prop-2-enoic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: QFQYZMGOKIROEC-RQOWECAXSA-N
SMILES: OC(=O)\C=C/C1=CC=C2OCOC2=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)