accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2373-89-9

IUPAC Name: (2E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
Synonyms: (2E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 268.31
Molecular Formula: C17H16O3
InChi Key: HDXVSZWKIHQDES-LFYBBSHMSA-N
SMILES: COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results