accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHO

CAS RN 23761-23-1

IUPAC Name: 3-oxocyclobutane-1-carboxylic acid
Synonyms: 3-oxocyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: IENOFRJPUPTEMI-UHFFFAOYSA-N
SMILES: OC(=O)C1CC(=O)C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)