accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23779-17-1

IUPAC Name: N-(4-iodophenyl)cyclopropanecarboxamide
Synonyms: N-(4-iodophenyl)cyclopropanecarboxamide
Molecular Weight (g/mol): 287.1
Molecular Formula: C10H10INO
InChi Key: OWRJMRAKRDWZHS-UHFFFAOYSA-N
SMILES: IC1=CC=C(NC(=O)C2CC2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results