CAS RN 23779-32-0
IUPAC Name:
[3-(triethoxysilyl)propyl]urea
Synonyms:
3-(triethoxysilyl)propylurea
Molecular Weight (g/mol):
264.4
Molecular Formula:
C10H24N2O4Si
InChi Key:
LVNLBBGBASVLLI-UHFFFAOYSA-N
SMILES:
CCO[Si](CCCNC(N)=O)(OCC)OCC
1
–
1
of
1
results
1
1-[3-(Triethoxysilyl)propyl]urea (40-52% in Methanol), TCI America™
CAS: 23779-32-0 Molecular Formula: C10H24N2O4Si Molecular Weight (g/mol): 264.397 MDL Number: MFCD00047877 InChI Key: LVNLBBGBASVLLI-UHFFFAOYSA-N Synonym: Triethoxy-3-ureidopropylsilane, 3-Ureidopropyltriethoxysilane PubChem CID: 90258 IUPAC Name: 3-triethoxysilylpropylurea SMILES: CCO[Si](CCCNC(=O)N)(OCC)OCC