accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23783-42-8

IUPAC Name: 2,5,8,11-tetraoxatridecan-13-ol
Synonyms: 2,5,8,11-tetraoxatridecan-13-ol
Molecular Weight (g/mol): 208.25
Molecular Formula: C9H20O5
InChi Key: ZNYRFEPBTVGZDN-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (4)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)