accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHN

CAS RN 23785-21-9

IUPAC Name: ethyl 1H-imidazole-5-carboxylate
Synonyms: ethyl 3H-imidazole-4-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: KLWYPRNPRNPORS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=CN1
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)