accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2379-60-4

IUPAC Name: 2,3-dichloro-6-nitroquinoxaline
Synonyms: 2,3-dichloro-6-nitroquinoxaline
Molecular Weight (g/mol): 244.03
Molecular Formula: C8H3Cl2N3O2
InChi Key: SFJCUOAQTGDBPO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=C(Cl)C(Cl)=NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results