accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23794-16-3

IUPAC Name: 2-bromo-1-(2-chloropyridin-4-yl)ethan-1-one
Synonyms: 2-bromo-1-(2-chloropyridin-4-yl)ethanone
Molecular Weight (g/mol): 234.48
Molecular Formula: C7H5BrClNO
InChi Key: BZWBRLWSWBQIBX-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results