accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHNN

CAS RN 23814-12-2

IUPAC Name: 2H-1,2,3-benzotriazole-5-carboxylic acid
Synonyms: 2H-1,2,3-benzotriazole-5-carboxylic acid
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: GUOVBFFLXKJFEE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=NNN=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results