accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOPOOHHOOPOOHOHHONaNaNaNaNa

CAS RN 2382-56-1

IUPAC Name: 2-{hydroxybis[4-(phosphonooxy)phenyl]methyl}benzoic acid pentasodium
Synonyms: 2-{hydroxybis[4-(phosphonooxy)phenyl]methyl}benzoic acid pentasodium
Molecular Weight (g/mol): 611.25
Molecular Formula: C20H18Na5O11P2
InChi Key: BETACOHSDKIQSV-UHFFFAOYSA-N
SMILES: [Na].[Na].[Na].[Na].[Na].OC(=O)C1=CC=CC=C1C(O)(C1=CC=C(OP(O)(O)=O)C=C1)C1=CC=C(OP(O)(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results