accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 238418-75-2

IUPAC Name: 4-(4-fluoro-2-nitrophenyl)morpholine
Synonyms: 4-(4-fluoro-2-nitrophenyl)morpholine
Molecular Weight (g/mol): 226.21
Molecular Formula: C10H11FN2O3
InChi Key: XEVFYVWDTJTEHQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(F)=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results