accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OClH3C

CAS RN 23844-56-6

IUPAC Name: methyl 2-(4-chloro-2-methylphenoxy)propanoate
Synonyms: methyl 2-(4-chloro-2-methylphenoxy)propanoate
Molecular Weight (g/mol): 228.67
Molecular Formula: C11H13ClO3
InChi Key: YWGAULPFWIQKRB-UHFFFAOYNA-N
SMILES: COC(=O)C(C)OC1=CC=C(Cl)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results