accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClClClClClCl

CAS RN 2385-85-5

IUPAC Name: dodecachloropentacyclo[5.3.0.0²,⁶.0³,⁹.0⁴,⁸]decane
Synonyms: mirex dechlorane dodecachloropentacyclo[5.2.1.02,6.03,9.05,8]decane (mirex) perchloropentacyclodecane
Molecular Weight (g/mol): 545.51
Molecular Formula: C10Cl12
InChi Key: GVYLCNUFSHDAAW-UHFFFAOYSA-N
SMILES: ClC12C3(Cl)C4(Cl)C5(Cl)C(Cl)(C1(Cl)C4(Cl)Cl)C2(Cl)C(Cl)(Cl)C35Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results