accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2386-26-7

IUPAC Name: ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: ALRDOFWBPAZOCW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C(C)=O)=C(C)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results