accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2386-52-9

IUPAC Name: methanesulfonic acid silver
Synonyms: methanesulfonic acid silver
Molecular Weight (g/mol): 203.97
Molecular Formula: CH4AgO3S
InChi Key: GTBQRHOYAUGRPV-UHFFFAOYSA-N
SMILES: [Ag].CS(O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)