accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOHO

CAS RN 23866-72-0

IUPAC Name: ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate
Synonyms: ethyl 7-hydroxy-4-oxochromene-2-carboxylate
Molecular Weight (g/mol): 234.21
Molecular Formula: C12H10O5
InChi Key: RKIMFICEWCXBCE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results