accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOCH3CH3Br

CAS RN 23877-12-5

IUPAC Name: tert-butyl 2-bromo-2-methylpropanoate
Synonyms: tert-butyl 2-bromo-2-methylpropanoate
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: IGVNJALYNQVQIT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C(C)(C)Br
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)