accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiH3CO

CAS RN 2388-07-0

IUPAC Name: lithium(1+) ethanolate
Synonyms: lithium(1+) ethoxide
Molecular Weight (g/mol): 52
Molecular Formula: C2H5LiO
InChi Key: AZVCGYPLLBEUNV-UHFFFAOYSA-N
SMILES: [Li+].CC[O-]
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)